3-Chloro-2-hydroxy-4-(trifluoromethyl)phenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

33223-1G 33223-25G 33223-500MG 33223-5G
76914-648EA 300.24 USD
76914-648 76914-650 76914-652 76914-654
3-Chloro-2-hydroxy-4-(trifluoromethyl)phenylboronic acid ≥95%
3-Chloro-2-hydroxy-4-(trifluoromethyl)phenylboronic acid
Formula: C₇H₅BClF₃O₃
MW: 240.372 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30834033
CAS Number: 2121512-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR