3,4-Difluoro-5-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

33181-1G 33181-25G 33181-500MG 33181-5G
76914-632EA 153.84 USD
76914-632 76914-634 76914-636 76914-638
3,4-Difluoro-5-methoxybenzoic acid ≥97%
3,4-Difluoro-5-methoxybenzoic acid
Formula: C₈H₆F₂O₃
MW: 188.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04972453
CAS Number: 887584-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR