3,4-Difluoro-5-methoxyphenol ≥97% research grade

Supplier: AOBChem USA

Warning

33178-1G 33178-25G 33178-500MG 33178-5G
76914-618EA 310.85 USD
76914-618 76914-620 76914-622 76914-624
3,4-Difluoro-5-methoxyphenol ≥97% research grade
3,4-Difluoro-5-methoxyphenol
Formula: C₇H₆F₂O₂
MW: 160.118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22414498
CAS Number: 1394915-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR