3,4-Dichloro-2-methoxybenzoic acid ≥95%
Supplier: AOBChem USA
|
Warning
|
33154-1G
33154-25G
33154-5G
76914-604EA
72.15
USD
76914-604
76914-606
76914-608
3,4-Dichloro-2-methoxybenzoic acid ≥95%
3,4-Dichloro-2-methoxybenzoic acid
Formula:
C₈H₆Cl₂O₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD28123955 CAS Number: 155382-86-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...