3-Chloro-4-methoxy-5-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

33122-1G 33122-25G 33122-500MG 33122-5G
76914-584EA 439.22 USD
76914-584 76914-586 76914-588 76914-590
3-Chloro-4-methoxy-5-(trifluoromethyl)benzoic acid ≥97%
3-Chloro-4-methoxy-5-(trifluoromethyl)benzoic acid
Formula: C₉H₆ClF₃O₃
MW: 254.59 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30723170
CAS Number: 1272528-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR