3-Bromo-5-nitrobenzonitrile ≥97%

Supplier: AOBChem USA

Warning

33103-1G 33103-25G 33103-5G
76914-574EA 72.15 USD
76914-574 76914-576 76914-578
3-Bromo-5-nitrobenzonitrile ≥97%
3-Bromo-5-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃BrN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00463804
CAS Number: 49674-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR