3-Bromo-2-(propan-2-yloxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

32964-1G 32964-25G 32964-500MG 32964-5G
76914-558EA 376.62 USD
76914-558 76914-560 76914-562 76914-564
3-Bromo-2-(propan-2-yloxy)benzaldehyde ≥97%
3-Bromo-2-(propan-2-yloxy)benzaldehyde
Formula: C₁₀H₁₁BrO₂
MW: 243.097 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14656660
CAS Number: 1247624-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR