3-Chloro-2-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

32952-25G 32952-5G
76914-508EA 346.92 USD
76914-508 76914-510
3-Chloro-2-methoxybenzaldehyde ≥97%
3-Chloro-2-methoxybenzaldehyde
Formula: C₈H₇ClO₂
MW: 170.5930 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09999135
CAS Number: 223778-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR