3-Bromo-2-iodobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

32856-1G 32856-25G 32856-5G
76914-482EA 84.88 USD
76914-482 76914-484 76914-486
3-Bromo-2-iodobenzoic acid ≥95%
3-Bromo-2-iodobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄BrIO₂
MW: 326.914 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15527220
CAS Number: 503821-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR