3-Fluoro-4,5-dimethoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

32775-1G 32775-25G 32775-500MG 32775-5G
76914-466EA 354.35 USD
76914-466 76914-468 76914-470 76914-472
3-Fluoro-4,5-dimethoxybenzaldehyde ≥97%
3-Fluoro-4,5-dimethoxybenzaldehyde
Formula: C₉H₉FO₃
MW: 184.164 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12756549
CAS Number: 71924-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR