3-Chloro-5-fluoro-4-hydroxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

32658-1G 32658-25G 32658-500MG 32658-5G
76914-418EA 439.22 USD
76914-418 76914-420 76914-422 76914-424
3-Chloro-5-fluoro-4-hydroxybenzoic acid ≥97%
3-Chloro-5-fluoro-4-hydroxybenzoic acid
Formula: C₇H₄ClFO₃
Storage Temperature: Refrigerator
MDL Number: MFCD20641557
CAS Number: 455-57-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR