2-(4-(Benzyloxy)-3-chloro-5-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Benzyloxy-3-chloro-5-fluorobenzeneboronic acid pinacol ester

32650-1G 32650-25G 32650-500MG 32650-5G
76914-402EA 153.84 USD
76914-402 76914-404 76914-406 76914-408
2-(4-(Benzyloxy)-3-chloro-5-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(4-(Benzyloxy)-3-chloro-5-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₉H₂₁BClFO₃
MW: 362.6307 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30529839
CAS Number: 2121515-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR