3-Chloro-5-fluoro-4-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

32641-25G 32641-5G
76914-392EA 559.1 USD
76914-392 76914-394
3-Chloro-5-fluoro-4-hydroxybenzaldehyde ≥97%
3-Chloro-5-fluoro-4-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄ClFO₂
Storage Temperature: Refrigerator
CAS Number: 870704-13-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR