(3-Chloro-4-ethoxy-5-fluorophenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloro-4-ethoxy-5-fluorophenylboronic acid

32638-1G 32638-25G 32638-500MG 32638-5G
76914-376EA 144.29 USD
76914-376 76914-378 76914-380 76914-382
(3-Chloro-4-ethoxy-5-fluorophenyl)boronic acid ≥97% research grade
(3-Chloro-4-ethoxy-5-fluorophenyl)boronic acid
Formula: C₈H₉BClFO₃
MW: 218.41 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20441841
CAS Number: 2096334-30-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR