3-Bromoquinoline-1-oxide ≥97% research grade

Supplier: AOBChem USA

Warning

32617-25G 32617-5G
76914-334EA 559.1 USD
76914-334 76914-336
3-Bromoquinoline-1-oxide ≥97% research grade
3-Bromoquinoline-1-oxide
Formula: C₉H₆BrNO
Storage Temperature: Ambient
MDL Number: MFCD01646407
CAS Number: 22615-00-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR