3-Chloro-2-ethoxypyridine-4-boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

32608-1G 32608-25G 32608-500MG 32608-5G
76914-318EA 238.71 USD
76914-318 76914-320 76914-322 76914-324
3-Chloro-2-ethoxypyridine-4-boronic acid ≥97% research grade
3-Chloro-2-ethoxypyridine-4-boronic acid
Formula: C₇H₉BClNO₃
Storage Temperature: Ambient
MDL Number: MFCD18914508
CAS Number: 2096333-52-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR