3-Chloro-2-methoxyisonicotinic acid ≥97% research grade
Supplier: AOBChem USA
|
Warning
|
32600-1G
32600-25G
32600-500MG
32600-5G
76914-304EA
260.99
USD
76914-304
76914-306
76914-308
76914-310
3-Chloro-2-methoxyisonicotinic acid ≥97% research grade
3-Chloro-2-methoxyisonicotinic acid
Formula:
C₇H₆ClNO₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD13185829 CAS Number: 1211584-06-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...