3-Chloro-2-formyl-4-methylphenylboronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

32545-1G 32545-25G 32545-500MG 32545-5G
76914-290EA 201.58 USD
76914-290 76914-292 76914-294 76914-296
3-Chloro-2-formyl-4-methylphenylboronic acid pinacol ester ≥97% research grade
3-Chloro-2-formyl-4-methylphenylboronic acid pinacol ester
Formula: C₁₄H₁₈BClO₃
Storage Temperature: Refrigerator
MDL Number: MFCD30529823
CAS Number: 2121513-75-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR