4-Bromo-2,3-difluorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

32514-1G 32514-25G 32514-500MG 32514-5G
76914-278EA 320.4 USD
76914-278 76914-280 76914-282 76914-284
4-Bromo-2,3-difluorobenzyl alcohol ≥97%
4-Bromo-2,3-difluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₅BrF₂O
Storage Temperature: Refrigerator
MDL Number: MFCD22493515
CAS Number: 162744-55-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR