3-Chloro-5-methoxybenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
32491-25G
32491-5G
76914-266EA
219.61
USD
76914-266
76914-268
3-Chloro-5-methoxybenzoic acid ≥97%
3-Chloro-5-methoxybenzoic acid
Precaución: for Research Use Only
Formula:
C₈H₇ClO₃ Storage Temperature: Ambient |
MDL Number:
MFCD01317994 CAS Number: 82477-67-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...