3-Methoxy-5-methylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(3-Methoxy-5-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

32436-1G 32436-25G 32436-500MG 32436-5G
76914-250EA 334.19 USD
76914-250 76914-252 76914-254 76914-256
3-Methoxy-5-methylphenylboronic acid pinacol ester ≥97%
3-Methoxy-5-methylphenylboronic acid pinacol ester
Formula: C₁₄H₂₁BO₃
MW: 248.126 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22493759
CAS Number: 365564-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR