3-Methoxy-4-(trifluoromethoxy)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

32323-1G 32323-500MG 32323-5G
76914-222EA 765.97 USD
76914-222 76914-224 76914-226
3-Methoxy-4-(trifluoromethoxy)benzoic acid ≥97%
3-Methoxy-4-(trifluoromethoxy)benzoic acid
Formula: C₉H₇F₃O₄
MW: 236.145 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18398449
CAS Number: 1261652-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR