3-Bromo-2,6-dimethoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

32301-25G 32301-5G
76914-176EA 622.75 USD
76914-176 76914-178
3-Bromo-2,6-dimethoxyphenylboronic acid ≥97%
3-Bromo-2,6-dimethoxyphenylboronic acid
Formula: C₈H₁₀BBrO₄
MW: 260.878 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30480183
CAS Number: 2121515-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR