3-Ethyl-4-methoxybenzaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

32267-1G 32267-25G 32267-500MG 32267-5G
76914-156EA 428.61 USD
76914-156 76914-158 76914-160 76914-162
3-Ethyl-4-methoxybenzaldehyde ≥97% research grade
3-Ethyl-4-methoxybenzaldehyde
Formula: C₁₀H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD06247460
CAS Number: 103323-26-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR