3-Chloro-2-fluoro-4-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

32256-1G 32256-500MG 32256-5G
76914-142EA 635.48 USD
76914-142 76914-144 76914-146
3-Chloro-2-fluoro-4-methoxyphenol ≥97%
3-Chloro-2-fluoro-4-methoxyphenol
Formula: C₇H₆ClFO₂
MW: 176.573 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28519989
CAS Number: 1781486-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR