3-Methoxy-2-naphthaleneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Methoxynaphthalen-2-boronic acid, (3-Methoxynaphthalen-2-yl)boronic acid

32247-1G 32247-25G 32247-5G
76914-128EA 96.55 USD
76914-128 76914-130 76914-132
3-Methoxy-2-naphthaleneboronic acid ≥97%
3-Methoxy-2-naphthaleneboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₁BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD12025968
CAS Number: 104115-76-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR