3-Formyl-2-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

32107-1G 32107-25G 32107-500MG 32107-5G
76914-094EA 379.81 USD
76914-094 76914-096 76914-098 76914-100
3-Formyl-2-methylbenzoic acid ≥97%
3-Formyl-2-methylbenzoic acid
Formula: C₉H₈O₃
MW: 164.158 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22414487
CAS Number: 1289037-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR