3-(Hydroxymethyl)-2-methylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

32074-1G 32074-25G 32074-500MG 32074-5G
76914-080EA 350.1 USD
76914-080 76914-082 76914-084 76914-086
3-(Hydroxymethyl)-2-methylphenylboronic acid pinacol ester ≥97%
3-(Hydroxymethyl)-2-methylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₁BO₃
MW: 248.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18729960
CAS Number: 1400755-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR