3-Fluoro-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

31991-1G 31991-25G 31991-500MG 31991-5G
76914-066EA 206.88 USD
76914-066 76914-068 76914-070 76914-072
3-Fluoro-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde ≥97%
3-Fluoro-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
Formula: C₁₃H₁₆BFO₃
MW: 250.0800 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18383509
CAS Number: 1246633-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR