4,5-Dibromo-2-furoic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4,5-Dibromofuran-2-carboxylic acid

31916-25G 31916-5G
76914-050EA 183.54 USD
76914-050 76914-052
4,5-Dibromo-2-furoic acid ≥97%
4,5-Dibromo-2-furoic acid

Precaución: for Research Use Only

Formula: C₅H₂Br₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00092311
CAS Number: 2434-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR