4-Benzyloxy-3-ethylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

31763-1G 31763-500MG 31763-5G
76914-038EA 485.9 USD
76914-038 76914-040 76914-042
4-Benzyloxy-3-ethylphenylboronic acid ≥97%
4-Benzyloxy-3-ethylphenylboronic acid
Formula: C₁₅H₁₇BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD31805755
CAS Number: 2137973-94-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR