3-Ethyl-4-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

31652-1G 31652-25G 31652-500MG 31652-5G
76914-012EA 379.81 USD
76914-012 76914-014 76914-016 76914-018
3-Ethyl-4-methoxyphenylboronic acid ≥97%
3-Ethyl-4-methoxyphenylboronic acid
Formula: C₉H₁₃BO₃
MW: 180.009 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13659400
CAS Number: 947547-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR