3-(Benzyloxy)-6-bromo-2-fluorophenol ≥97%

Supplier: AOBChem USA

Warning

31487-1G 31487-25G 31487-5G
76913-990EA 96.55 USD
76913-990 76913-992 76913-994
3-(Benzyloxy)-6-bromo-2-fluorophenol ≥97%
3-(Benzyloxy)-6-bromo-2-fluorophenol

Precaución: for Research Use Only

Formula: C₁₃H₁₀BrFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD16618956
CAS Number: 1228956-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR