4,5-Dichloro-2-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

31230-1G 31230-25G 31230-500MG 31230-5G
76913-876EA 409.51 USD
76913-876 76913-878 76913-880 76913-882
4,5-Dichloro-2-fluorobenzaldehyde ≥97%
4,5-Dichloro-2-fluorobenzaldehyde
Formula: C₇H₃Cl₂FO
MW: 193.003 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28785955
CAS Number: 1217800-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR