2-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzenemethanol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Hydroxymethyl-4-methoxyphenylboronic acid pinacol ester

31137-1G 31137-25G 31137-5G
76913-818EA 84.88 USD
76913-818 76913-820 76913-822
2-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzenemethanol ≥97%
2-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzenemethanol
Formula: C₁₄H₂₁BO₄
MW: 264.12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13184694
CAS Number: 1009303-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR