2-(3-Chloro-2,4-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloro-2,4-difluorophenylboronic acid pinacol ester

31135-1G 31135-25G 31135-500MG 31135-5G
76913-806EA 142.17 USD
76913-806 76913-808 76913-810 76913-812
2-(3-Chloro-2,4-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(3-Chloro-2,4-difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₂H₁₄BClF₂O₂
MW: 274.49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29088265
CAS Number: 1799614-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR