4,5,6-Trichloronicotinic acid ≥97%

Supplier: AOBChem USA

Warning

31129-1G 31129-500MG 31129-5G
76913-794EA 498.63 USD
76913-794 76913-796 76913-798
4,5,6-Trichloronicotinic acid ≥97%
4,5,6-Trichloronicotinic acid
Formula: C₆H₂Cl₃NO₂
MW: 226.445 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11044391
CAS Number: 847608-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR