2-Methoxy-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3-Formyl-4-methoxyphenylboronic acid pinacol ester

31064-25G 31064-5G
76913-754EA 342.68 USD
76913-754 76913-756
2-Methoxy-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde ≥97% research grade
2-Methoxy-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
Formula: C₁₄H₁₉BO₄
MW: 262.10 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22494881
CAS Number: 866546-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR