3,5-Dichloro-2-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

31052-1G 31052-25G 31052-500MG 31052-5G
76913-738EA 260.99 USD
76913-738 76913-740 76913-742 76913-744
3,5-Dichloro-2-fluorobenzaldehyde ≥97%
3,5-Dichloro-2-fluorobenzaldehyde
Formula: C₇H₃Cl₂FO
MW: 193.003 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26687657
CAS Number: 477535-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR