3-Chloro-4-propoxyphenylboronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

31015-25G 31015-5G
76913-718EA 622.75 USD
76913-718 76913-720
3-Chloro-4-propoxyphenylboronic acid pinacol ester ≥95%
3-Chloro-4-propoxyphenylboronic acid pinacol ester
Formula: C₁₅H₂₂BClO₃
Storage Temperature: Ambient
MDL Number: MFCD29088247
CAS Number: 1688655-72-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR