3,4-Difluoro-2-methylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(3,4-Difluoro-2-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

30945-1G 30945-25G 30945-5G
76913-710EA 89.12 USD
76913-710 76913-712 76913-714
3,4-Difluoro-2-methylphenylboronic acid pinacol ester ≥97%
3,4-Difluoro-2-methylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BF₂O₂
MW: 254.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08669570
CAS Number: 1025707-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR