3-Acetyl-2,6-dimethoxypyridine ≥97%

Supplier: AOBChem USA

Warning

30920-1G 30920-25G 30920-5G
76913-670EA 62.6 USD
76913-670 76913-672 76913-674
3-Acetyl-2,6-dimethoxypyridine ≥97%
3-Acetyl-2,6-dimethoxypyridine

Precaución: for Research Use Only

Formula: C₉H₁₁NO₃
MW: 181.189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07784407
CAS Number: 870703-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR