3-Bromo-2-chloro-6-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

30911-25G 30911-5G
76913-660EA 257.8 USD
76913-660 76913-662
3-Bromo-2-chloro-6-methoxypyridine ≥97%
3-Bromo-2-chloro-6-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅BrClNO
Storage Temperature: Refrigerator
MDL Number: MFCD10699710
CAS Number: 777931-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR