3-Bromo-5-iodo-2-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

30715-1G 30715-25G 30715-500MG 30715-5G
76913-608EA 200.52 USD
76913-608 76913-610 76913-612 76913-614
3-Bromo-5-iodo-2-methylbenzoic acid ≥95%
3-Bromo-5-iodo-2-methylbenzoic acid
Formula: C₈H₆BrIO₂
MW: 340.941 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20526668
CAS Number: 1022983-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR