3-(4-Methoxyphenyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

30279-1G 30279-25G 30279-500MG 30279-5G
76913-522EA 292.81 USD
76913-522 76913-524 76913-526 76913-528
3-(4-Methoxyphenyl)pyridine ≥95%
3-(4-Methoxyphenyl)pyridine
Formula: C₁₂H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD06801825
CAS Number: 5958-02-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR