3-(Benzyloxy)-2,6-difluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

30224-1G 30224-500MG 30224-5G
76913-512EA 468.92 USD
76913-512 76913-514 76913-516
3-(Benzyloxy)-2,6-difluorobenzoic acid ≥97%
3-(Benzyloxy)-2,6-difluorobenzoic acid
Formula: C₁₄H₁₀F₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD22392201
CAS Number: 1421604-28-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR