3-Bromo-5-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

29809-1G 29809-25G 29809-500MG 29809-5G
76913-416EA 129.43 USD
76913-416 76913-418 76913-420 76913-422
3-Bromo-5-formylbenzoic acid ≥97%
3-Bromo-5-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₅BrO₃
MW: 229.027 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22544371
CAS Number: 398119-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR