4-(1,3,2-Dioxaborinan-2-yl)phenol ≥97%

Supplier: AOBChem USA

Warning

29778-25G 29778-5G
76913-406EA 215.37 USD
76913-406 76913-408
4-(1,3,2-Dioxaborinan-2-yl)phenol ≥97%
4-(1,3,2-Dioxaborinan-2-yl)phenol

Precaución: for Research Use Only

Formula: C₉H₁₁BO₃
MW: 177.993 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21609692
CAS Number: 1640035-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR