2-Bromo-4,5-diluorotoluene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4,5-Difluoro-2-methylbromobenzene, 1-Bromo-3,4-difluoro-6-methylbenzene, 1-Bromo-4,5-difluoro-2-methylbenzene

29748-25G 29748-5G
76913-396EA 187.78 USD
76913-396 76913-398
2-Bromo-4,5-diluorotoluene ≥97%
2-Bromo-4,5-diluorotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrF₂
MW: 207.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07777160
CAS Number: 875664-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR