3-Methyl-4-(Methylsulfanyl)Benzoic Acid ≥97%
Supplier: AOBChem USA
|
Warning
|
29743-1G
29743-25G
29743-500MG
29743-5G
76913-382EA
260.99
USD
76913-382
76913-384
76913-386
76913-388
3-Methyl-4-(Methylsulfanyl)Benzoic Acid ≥97%
3-Methyl-4-(Methylsulfanyl)Benzoic Acid
Formula:
C₉H₁₀O₂S Storage Temperature: Refrigerator |
MDL Number:
MFCD16853040 CAS Number: 851669-34-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...